siRNA Details
Gene Symbol: STEAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008410.1 | NP_001008410.1 | 5 | 1624 | ||
NM_018234.2 | NP_060704.2 | 6 | 1692 | |||
NM_138637.2 | NP_619543.2 | 6 | 1856 | |||
NM_182915.2 | NP_878919.2 | 6 | 2039 | |||
XM_006712614.3 | XP_006712677.1 | 6 | 1766 | |||
XM_006712615.1 | XP_006712678.1 | 5 | 1636 | |||
XM_011511403.1 | XP_011509705.1 | 7 | 2050 | |||
GenBank mRNA | AF238864.1 | - | 5 | 1580 | ||
AF262322.1 | - | 6 | 1812 | |||
AF423424.1 | - | 6 | 1669 | |||
AK001691.1 | - | 6 | 1669 | |||
AK024163.1 | - | 4 | 583 | |||
AK291608.1 | - | 6 | 2001 | |||
AL833624.1 | - | 6 | 2039 | |||
AY029585.1 | - | 6 | 1669 | |||
AY082673.1 | - | 5 | 1576 | |||
AY214461.1 | - | 6 | 1669 | |||
BC042150.1 | - | 5 | 1575 | 4586693 | ||
BC095421.1 | - | 5 | 1579 | 30336214 | ||
BX538047.1 | - | 6 | 2008 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
