Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPANXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032417.3 | NP_115793.1 | 2 | 450 | ||
GenBank mRNA | AF312765.1 | - | 2 | 126 | ||
AJ457795.1 | - | 2 | 152 | |||
AY920734.1 | - | 2 | 116 | |||
AY920740.1 | - | 2 | 116 | |||
BC005382.1 | - | 2 | 209 | 3935644 | ||
BC062672.1 | - | 2 | 390 | 4047937 | ||
BC103961.1 | - | 2 | 149 | 40013988 | ||
BC121070.2 | - | 2 | 159 | 40119569 | ||
BC134418.1 | - | 2 | 159 | 40119568 | ||
BM554791.1 | - | 2 | 450 |
Gene Symbol: SPANXC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022661.3 | NP_073152.2 | 2 | 209 |