Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SNAP91 SIRNA Assays ›
Gene Symbol:
SNAP91
Gene Name:
synaptosome associated protein 91
Gene Aliases:
AP180, CALM
Chromosome Location:
Chr.6: 83552885 - 83709408 on Build GRCh38
UniGene ID:
Hs.368046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
225032
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SNAP91
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242792.1
NP_001229721.1
19
2076
NM_001242793.1
NP_001229722.1
18
1869
NM_001256718.1
NP_001243647.1
18
1786
NM_014841.2
NP_055656.1
19
2079
XM_005248770.4
XP_005248827.1
19
1918
XM_006715615.1
XP_006715678.1
19
1870
XM_011536265.1
XP_011534567.1
19
1877
XM_011536266.1
XP_011534568.1
19
1989
XM_011536269.1
XP_011534571.1
19
1870
XM_011536271.1
XP_011534573.1
18
1828
XM_011536275.2
XP_011534577.1
19
1870
XM_011536276.2
XP_011534578.1
18
1762
XM_011536277.1
XP_011534579.1
21
2158
XM_017011554.1
XP_016867043.1
18
2028
XM_017011555.1
XP_016867044.1
18
1912
XM_017011556.1
XP_016867045.1
18
1844
XM_017011557.1
XP_016867046.1
19
2047
XM_017011558.1
XP_016867047.1
19
1877
XM_017011559.1
XP_016867048.1
19
1976
XM_017011560.1
XP_016867049.1
19
1918
XM_017011561.1
XP_016867050.1
19
1989
XM_017011562.1
XP_016867051.1
18
2032
XM_017011563.1
XP_016867052.1
17
2026
XM_017011564.1
XP_016867053.1
17
1897
XM_017011565.1
XP_016867054.1
17
1955
XM_017011566.1
XP_016867055.1
18
2005
XM_017011567.1
XP_016867056.1
18
1808
XM_017011568.1
XP_016867057.1
17
1999
XM_017011569.1
XP_016867058.1
18
2005
XM_017011574.1
XP_016867063.1
19
2047
XM_017011575.1
XP_016867064.1
19
2047
XM_017011576.1
XP_016867065.1
19
1850
XM_017011577.1
XP_016867066.1
19
1976
XM_017011578.1
XP_016867067.1
19
1989
XM_017011579.1
XP_016867068.1
19
2047
XM_017011580.1
XP_016867069.1
18
1939
XM_017011581.1
XP_016867070.1
18
1769
XM_017011582.1
XP_016867071.1
18
1798
XM_017011583.1
XP_016867072.1
18
1926
XM_017011584.1
XP_016867073.1
18
1769
XM_017011585.1
XP_016867074.1
18
1810
XM_017011586.1
XP_016867075.1
18
1762
XM_017011587.1
XP_016867076.1
18
1868
XM_017011588.1
XP_016867077.1
18
1881
XM_017011589.1
XP_016867078.1
17
1897
XM_017011590.1
XP_016867079.1
18
1810
GenBank mRNA
AB014556.1
-
19
2005
AK289582.1
-
19
2047
AK294556.1
-
12
1037
AK294898.1
-
18
1770
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -
vesicle coat protein
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
protein transport
clathrin coat assembly
regulation of clathrin-mediated endocytosis
protein binding
1-phosphatidylinositol binding
protein kinase binding
clathrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-p89b7:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0