Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAV20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127936.1 | - | 3 | 690 |
Gene Symbol: OR6C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M87866.1 | - | 148 |