Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015258.1 | NP_056073.1 | 21 | 2232 | ||
XM_006717018.2 | XP_006717081.1 | 21 | 2119 | |||
XM_006717019.1 | XP_006717082.1 | 20 | 2080 | |||
XM_017014552.1 | XP_016870041.1 | 20 | 2050 | |||
GenBank mRNA | AB014574.1 | - | 21 | 2159 | ||
AK301208.1 | - | 8 | 986 | |||
AK303074.1 | - | 20 | 2080 | |||
BC077732.1 | - | 5 | 445 |