siRNA Details
Gene Symbol: DNHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144666.2 | NP_653267.2 | 21 | 7054 | ||
GenBank mRNA | AB290171.1 | - | 8 | 3754 | ||
AK074178.1 | - | 8 | 3725 | |||
AK125885.1 | - | 3 | 3214 | |||
AL137619.1 | - | 1 | 1399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
