Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033602.2 | NP_001028774.2 | 1 | 937 | ||
XM_011535019.2 | XP_011533321.1 | 3 | 1462 | |||
XM_011535020.2 | XP_011533322.1 | 3 | 1462 | |||
XM_011535021.2 | XP_011533323.1 | 3 | 1462 | |||
XM_017020500.1 | XP_016875989.1 | 3 | 1462 | |||
XM_017020501.1 | XP_016875990.1 | 3 | 1462 | |||
XM_017020502.1 | XP_016875991.1 | 3 | 1379 | |||
GenBank mRNA | AB018317.2 | - | 1 | 937 | ||
BC150245.1 | - | 1 | 958 | 9007469 |