Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207315.3 | NP_997198.2 | 5 | 1462 | ||
NR_046236.1 | 5 | 874 | ||||
GenBank mRNA | BC016969.1 | - | 2 | 166 | ||
DA501203.1 | - | 5 | 458 |