Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276252.1 | NP_001263181.1 | 16 | 3048 | ||
NM_015023.4 | NP_055838.2 | 16 | 3045 | |||
XM_011541057.1 | XP_011539359.1 | 16 | 2726 | |||
GenBank mRNA | AB028960.1 | - | 10 | 3391 | ||
AK001734.1 | - | 15 | 2462 | |||
AK023101.1 | - | 7 | 1587 | |||
AK023778.1 | - | 7 | 1627 | |||
BC032523.1 | - | 16 | 2908 | 5500384 |