Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002334.3 | NP_002325.2 | 13 | 1808 | ||
XM_011520103.2 | XP_011518405.1 | 7 | 878 | |||
XM_017017734.1 | XP_016873223.1 | 13 | 1725 | |||
GenBank mRNA | AB011540.1 | - | 5 | 578 | ||
AB084910.1 | - | 13 | 1808 | |||
AB231861.1 | - | 13 | 1711 | |||
BC136667.1 | - | 13 | 1729 | 9020657 | ||
BC136668.1 | - | 13 | 1729 | 9020658 |