Search
Search

TTLL12| Gene Symbol: TTLL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015140.3 | NP_055955.1 | 14 | 2858 | ||
| GenBank mRNA | AK123789.1 | - | 3 | 1928 | ||
| AK304464.1 | - | 4 | 1344 | |||
| BC001070.2 | - | 14 | 2822 | 3504490 | ||
| D63487.1 | - | 14 | 2870 | |||