Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PM20D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010853.2 | NP_001010853.1 | 6 | 1158 | ||
XM_005248661.2 | XP_005248718.1 | 7 | 1157 | |||
XM_005248662.4 | XP_005248719.1 | 7 | 908 | |||
XM_011535481.2 | XP_011533783.1 | 6 | 1390 | |||
XM_017010290.1 | XP_016865779.1 | 6 | 980 | |||
XM_017010291.1 | XP_016865780.1 | 7 | 800 | |||
GenBank mRNA | AK127529.1 | - | 4 | 453 | ||
AK302464.1 | - | 6 | 1118 | |||
BC035036.1 | - | 6 | 1161 | 5262128 | ||
BC045583.1 | - | 6 | 909 |