siRNA Details
Gene Symbol: NOTCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017617.4 | NP_060087.3 | 34 | 8897 | ||
XM_011518717.2 | XP_011517019.2 | 31 | 8189 | |||
GenBank mRNA | AB209873.1 | - | 26 | 7434 | ||
AK000012.1 | - | 1 | 1073 | |||
BC013208.1 | - | 4 | 3082 | |||
BC046127.1 | - | 1 | 648 | |||
CR457221.1 | - | 1 | 400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -




