Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009185.2 | NP_001009185.1 | 4 | 527 | ||
NM_001205247.1 | NP_001192176.1 | 4 | 422 | |||
NM_001205248.1 | NP_001192177.1 | 4 | 514 | |||
NM_001205250.1 | NP_001192179.1 | 4 | 524 | |||
NM_001205251.1 | NP_001192180.1 | 4 | 409 | |||
NM_015256.3 | NP_056071.2 | 4 | 527 | |||
GenBank mRNA | AB020644.1 | - | 4 | 487 | ||
AF102845.1 | - | 3 | 304 | |||
AF129166.1 | - | 4 | 457 | |||
AK294291.1 | - | 4 | 397 | |||
AK314452.1 | - | 4 | 429 | |||
BC026161.1 | - | 4 | 423 | 4830357 | ||
BC047453.1 | - | 4 | 331 | 5270879 | ||
BI464352.1 | - | 4 | 510 | |||
BI545119.1 | - | 4 | 528 | |||
BI548729.1 | - | 4 | 527 | |||
DQ083031.1 | - | 3 | 343 | |||
DQ655980.2 | - | 4 | 459 |