Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRIM37 SIRNA Assays ›
Gene Symbol:
TRIM37
Gene Name:
tripartite motif containing 37
Gene Aliases:
MUL, POB1, TEF3
Chromosome Location:
Chr.17: 58968068 - 59106921 on Build GRCh38
UniGene ID:
Hs.579079
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22668
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRIM37
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005207.3
NP_001005207.1
4, 5
744
NM_001320987.1
NP_001307916.1
3, 4
642
NM_001320988.1
NP_001307917.1
4, 5
744
NM_001320989.1
NP_001307918.1
4, 5
744
NM_001320990.1
NP_001307919.1
5, 6
820
NM_015294.4
NP_056109.1
4, 5
744
XM_005257385.1
XP_005257442.1
4, 5
576
XM_011524831.1
XP_011523133.1
4, 5
576
XM_011524832.2
XP_011523134.1
4, 5
576
XM_011524833.1
XP_011523135.1
4, 5
576
XM_011524834.1
XP_011523136.1
3, 4
458
XM_011524836.1
XP_011523138.1
4, 5
576
XM_017024662.1
XP_016880151.1
4, 5
576
XM_017024663.1
XP_016880152.1
4, 5
576
XM_017024664.1
XP_016880153.1
4, 5
576
XM_017024665.1
XP_016880154.1
3, 4
458
XM_017024666.1
XP_016880155.1
4, 5
576
XM_017024667.1
XP_016880156.1
4, 5
576
XM_017024668.1
XP_016880157.1
3, 4
458
XM_017024669.1
XP_016880158.1
3, 4
458
XM_017024670.1
XP_016880159.1
3, 4
457
XM_017024673.1
XP_016880162.1
3, 4
263
XM_017024674.1
XP_016880163.1
3, 4
263
GenBank mRNA
AB020705.1
-
4, 5
329
AF213365.1
-
4, 5
548
AK289674.1
-
4, 5
559
AK292459.1
-
4, 5
616
AK294850.1
-
3, 4
547
BC036012.1
-
4, 5
580
5270430
BX537955.1
-
5, 6
639
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
negative regulation of NF-kappaB transcription factor activity
histone H2A monoubiquitination
histone H2A-K119 monoubiquitination
negative regulation of centriole replication
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
protein autoubiquitination
aggresome assembly
chromatin binding
ubiquitin-protein transferase activity
tumor necrosis factor receptor binding
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase binding
protein homodimerization activity
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-grtsk:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0