Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164232.1 | NP_001157704.1 | 13 | 1760 | ||
NM_015214.2 | NP_056029.2 | 13 | 1937 | |||
XM_005273454.2 | XP_005273511.1 | 13 | 2085 | |||
XM_005273455.3 | XP_005273512.1 | 13 | 2085 | |||
XM_005273456.3 | XP_005273513.1 | 12 | 1995 | |||
XM_011544456.1 | XP_011542758.1 | 13 | 2085 | |||
XM_017013252.1 | XP_016868741.1 | 13 | 1894 | |||
XM_017013253.1 | XP_016868742.1 | 13 | 2085 | |||
XM_017013254.1 | XP_016868743.1 | 12 | 1674 | |||
XM_017013255.1 | XP_016868744.1 | 11 | 1683 | |||
XM_017013256.1 | XP_016868745.1 | 12 | 1773 | |||
GenBank mRNA | AB018268.1 | - | 10 | 1147 | ||
AK023218.1 | - | 9 | 966 | |||
AK056525.1 | - | 13 | 1839 | |||
AK094951.1 | - | 2 | 2350 | |||
AK125904.1 | - | 13 | 1760 | |||
AK127040.1 | - | 4 | 1288 | |||
AK299875.1 | - | 11 | 1344 | |||
BC010504.1 | - | 8 | 908 | 3451888 |