siRNA Details
Gene Symbol: PRRC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013318.3 | NP_037450.2 | 30, 31 | 6609 | ||
GenBank mRNA | AB011087.1 | - | 14, 15 | 1878 | ||
AK026162.1 | - | 7, 8 | 912 | |||
AK304849.1 | - | 3, 4 | 346 | |||
BC012289.1 | - | 8, 9 | 1101 | |||
BX647528.1 | - | 17, 18 | 4248 | |||
CR749818.1 | - | 25, 26 | 5684 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
