siRNA Details
Gene Symbol: PRRC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013318.3 | NP_037450.2 | 31 | 6627 | ||
GenBank mRNA | AB011087.1 | - | 15 | 1896 | ||
AK025675.1 | - | 3 | 221 | |||
AK026162.1 | - | 8 | 930 | |||
AK055916.1 | - | 3 | 925 | |||
AK303507.1 | - | 5 | 526 | |||
AK304849.1 | - | 4 | 364 | |||
AY700780.1 | - | 7 | 781 | |||
BC012289.1 | - | 9 | 1119 | |||
BX647528.1 | - | 18 | 4266 | |||
BX647842.1 | - | 17 | 2330 | |||
CR749818.1 | - | 26 | 5702 | |||
CR933661.1 | - | 18 | 4431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
