siRNA Details
Gene Symbol: PRRC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013318.3 | NP_037450.2 | 31 | 7328 | ||
GenBank mRNA | AB011087.1 | - | 15 | 2597 | ||
AK025675.1 | - | 3 | 922 | |||
AK026025.1 | - | 1 | 161 | |||
AK026162.1 | - | 9 | 1406 | |||
AK055916.1 | - | 3 | 1624 | |||
AK304849.1 | - | 4 | 1065 | |||
AL050142.1 | - | 1 | 100 | |||
BX647528.1 | - | 18 | 4967 | |||
BX647842.1 | - | 17 | 3031 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
