Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138466.1 | NP_612475.1 | 2, 3 | 202 | ||
GenBank mRNA | BC012365.1 | - | 1, 2 | 64 | 4576341 | |
BX374687.2 | - | 2, 3 | 206 |