Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278510.2 | NP_001265439.2 | 6 | 1166 | ||
NM_014518.3 | NP_055333.3 | 6 | 1295 | |||
NR_103551.2 | 6 | 1750 | ||||
XM_006723372.3 | XP_006723435.1 | 6 | 1369 | |||
XM_011527292.1 | XP_011525594.1 | 6 | 1277 | |||
XM_017027280.1 | XP_016882769.1 | 6 | 1824 | |||
GenBank mRNA | AB209357.1 | - | 6 | 1719 | ||
AF192979.1 | - | 6 | 1296 | |||
BC150612.1 | - | 4 | 1031 | 9056982 | ||
CR936724.1 | - | 6 | 1167 |