siRNA Details
Gene Symbol: PRRC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013318.3 | NP_037450.2 | 1, 2 | 167 | ||
GenBank mRNA | BC002872.2 | - | 2, 3 | 218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
