Search
Search

BICD2| Gene Symbol: BICD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003800.1 | NP_001003800.1 | 1, 2 | 302 | ||
| NM_015250.3 | NP_056065.1 | 1, 2 | 302 | |||
| GenBank mRNA | AB014599.1 | - | 1, 2 | 222 | ||
| AY052562.1 | - | 1, 2 | 302 | |||
| BC073970.1 | - | 1, 2 | 317 | 6169195 | ||