Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152432.2 | NP_689645.2 | 18 | 1697 | ||
XM_011542615.2 | XP_011540917.1 | 18 | 4185 | |||
XM_011542616.2 | XP_011540918.1 | 18 | 1954 | |||
XM_011542617.2 | XP_011540919.1 | 19 | 1840 | |||
XM_017017237.1 | XP_016872726.1 | 18 | 2143 | |||
XM_017017238.1 | XP_016872727.1 | 19 | 4326 | |||
GenBank mRNA | AK057372.1 | - | 6 | 567 | ||
BU146672.1 | - | 2 | 203 |