Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198578.3 | NP_940980.3 | 27 | 3850 | ||
XM_005268629.3 | XP_005268686.1 | 27 | 3966 | |||
XM_011537877.2 | XP_011536179.1 | 27 | 3966 | |||
XM_011537881.2 | XP_011536183.1 | 27 | 3966 | |||
XM_011537882.2 | XP_011536184.1 | 27 | 3966 | |||
XM_017018786.1 | XP_016874275.1 | 27 | 3966 | |||
XM_017018787.1 | XP_016874276.1 | 6 | 794 | |||
XM_017018789.1 | XP_016874278.1 | 27 | 3966 | |||
GenBank mRNA | AK127729.1 | - | 18 | 2580 | ||
AK131537.1 | - | 2 | 154 | |||
AL832453.2 | - | 6 | 1025 | |||
AY792511.1 | - | 27 | 3729 | |||
BC117180.1 | - | 27 | 3787 | 40125731 |