Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other R3HDM1 SIRNA Assays ›
Gene Symbol:
R3HDM1
Gene Name:
R3H domain containing 1
Gene Aliases:
R3HDM
Chromosome Location:
Chr.2: 135531429 - 135725270 on Build GRCh38
UniGene ID:
Hs.412462
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22790
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: R3HDM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282798.1
NP_001269727.1
6
822
NM_001282799.1
NP_001269728.1
4
685
NM_001282800.1
NP_001269729.1
3
479
NM_015361.3
NP_056176.2
6
817
XM_005263626.3
XP_005263683.1
6
806
XM_005263627.3
XP_005263684.1
6
806
XM_005263628.3
XP_005263685.1
6
806
XM_005263629.3
XP_005263686.1
6
806
XM_006712389.2
XP_006712452.1
6
806
XM_011510870.1
XP_011509172.1
6
806
XM_011510871.1
XP_011509173.1
6
822
XM_011510872.2
XP_011509174.1
5
764
XM_011510873.2
XP_011509175.1
6
806
XM_011510874.2
XP_011509176.1
4
674
XM_011510875.2
XP_011509177.1
4
690
XM_011510876.2
XP_011509178.1
6
806
XM_011510877.2
XP_011509179.1
6
806
XM_011510878.2
XP_011509180.1
6
806
XM_011510879.2
XP_011509181.1
6
806
XM_011510881.1
XP_011509183.1
6
806
XM_017003718.1
XP_016859207.1
6
822
XM_017003719.1
XP_016859208.1
5
801
XM_017003720.1
XP_016859209.1
6
822
XM_017003721.1
XP_016859210.1
6
822
XM_017003722.1
XP_016859211.1
4
674
XM_017003723.1
XP_016859212.1
4
690
XM_017003724.1
XP_016859213.1
5
764
XM_017003725.1
XP_016859214.1
7
898
XM_017003726.1
XP_016859215.1
7
968
XM_017003727.1
XP_016859216.1
6
926
XM_017003728.1
XP_016859217.1
6
806
XM_017003729.1
XP_016859218.1
6
822
XM_017003730.1
XP_016859219.1
4
566
XM_017003731.1
XP_016859220.1
7
919
XM_017003732.1
XP_016859221.1
7
897
XM_017003733.1
XP_016859222.1
6
808
XM_017003734.1
XP_016859223.1
7
968
XM_017003735.1
XP_016859224.1
4
566
XM_017003736.1
XP_016859225.1
7
965
GenBank mRNA
AK127793.1
-
3
450
AK290015.1
-
6
708
BC041093.1
-
6
759
5556247
D21852.1
-
4
639
Back To Top
More Information
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-vsgnr:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0