Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NEDD4L SIRNA Assays ›
Gene Symbol:
NEDD4L
Gene Name:
neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase
Gene Aliases:
NEDD4-2, NEDD4.2, RSP5, hNEDD4-2
Chromosome Location:
Chr.18: 58044362 - 58401540 on Build GRCh38
UniGene ID:
Hs.185677
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22855
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NEDD4L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144964.1
NP_001138436.1
31
4217
NM_001144965.1
NP_001138437.1
31
4169
NM_001144966.2
NP_001138438.1
30
4094
NM_001144967.2
NP_001138439.1
31
4263
NM_001144968.1
NP_001138440.1
31
4159
NM_001144969.1
NP_001138441.1
30
4099
NM_001144970.2
NP_001138442.1
29
4142
NM_001144971.1
NP_001138443.1
31
4187
NM_001243960.1
NP_001230889.1
29
4071
NM_015277.5
NP_056092.2
30
4203
XM_005266658.4
XP_005266715.2
28
4194
XM_005266660.4
XP_005266717.2
27
4138
XM_005266663.4
XP_005266720.2
26
4078
XM_006722421.3
XP_006722484.2
27
4133
XM_006722424.3
XP_006722487.2
24
3686
XM_006722425.3
XP_006722488.2
23
3630
XM_006722426.3
XP_006722489.1
33
4411
XM_006722428.3
XP_006722491.1
32
4333
XM_006722430.3
XP_006722493.1
31
4241
XM_011525887.2
XP_011524189.1
33
4391
XM_017025676.1
XP_016881165.1
25
3883
XM_017025677.1
XP_016881166.1
24
3827
XM_017025678.1
XP_016881167.1
31
4272
XM_017025679.1
XP_016881168.1
30
4187
XM_017025680.1
XP_016881169.1
27
3875
XM_017025681.1
XP_016881170.1
26
3677
GenBank mRNA
AB007899.1
-
29
4043
AB071179.1
-
30
4035
AF070601.1
-
1
788
AK091189.1
-
3
1911
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
protein polyubiquitination
regulation of membrane depolarization
protein monoubiquitination
sodium ion transport
cellular sodium ion homeostasis
excretion
response to metal ion
protein ubiquitination
viral life cycle
water homeostasis
ion transmembrane transport
regulation of ion transmembrane transport
regulation of protein catabolic process
regulation of membrane potential
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of protein catabolic process
positive regulation of endocytosis
regulation of membrane repolarization
protein K48-linked ubiquitination
ventricular cardiac muscle cell action potential
regulation of potassium ion transmembrane transporter activity
negative regulation of potassium ion transmembrane transporter activity
negative regulation of potassium ion transmembrane transport
negative regulation of sodium ion transmembrane transport
positive regulation of dendrite extension
negative regulation of protein localization to cell surface
negative regulation of sodium ion transmembrane transporter activity
positive regulation of caveolin-mediated endocytosis
ubiquitin-protein transferase activity
protein binding
potassium channel regulator activity
ligase activity
sodium channel regulator activity
potassium channel inhibitor activity
sodium channel inhibitor activity
ion channel binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kqhf4:80/10.239.162.79:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0