Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRIM37 SIRNA Assays ›
          Gene Symbol:
          TRIM37
          Gene Name:
          tripartite motif containing 37
          Gene Aliases:
          MUL, POB1, TEF3
          Chromosome Location:
          Chr.17: 58968068 - 59106921 on Build GRCh38
          UniGene ID:
          Hs.579079
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 22859

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TRIM37
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001005207.3 NP_001005207.1 13 1630
          NM_001320987.1 NP_001307916.1 12 1528
          NM_001320988.1 NP_001307917.1 13 1630
          NM_001320989.1 NP_001307918.1 13 1630
          NM_001320990.1 NP_001307919.1 14 1706
          NM_015294.4 NP_056109.1 13 1630
          XM_005257385.1 XP_005257442.1 13 1462
          XM_011524831.1 XP_011523133.1 13 1462
          XM_011524832.2 XP_011523134.1 13 1462
          XM_011524833.1 XP_011523135.1 12 1411
          XM_011524834.1 XP_011523136.1 12 1344
          XM_011524836.1 XP_011523138.1 13 1462
          XM_017024662.1 XP_016880151.1 13 1462
          XM_017024663.1 XP_016880152.1 13 1462
          XM_017024664.1 XP_016880153.1 12 1411
          XM_017024665.1 XP_016880154.1 12 1344
          XM_017024666.1 XP_016880155.1 12 1411
          XM_017024667.1 XP_016880156.1 12 1411
          XM_017024668.1 XP_016880157.1 12 1344
          XM_017024669.1 XP_016880158.1 11 1293
          XM_017024670.1 XP_016880159.1 12 1343
          XM_017024671.1 XP_016880160.1 12 1374
          XM_017024672.1 XP_016880161.1 12 1374
          XM_017024673.1 XP_016880162.1 12 1166
          XM_017024674.1 XP_016880163.1 12 1166
          GenBank mRNA AB020705.1 - 13 1215
          AF213365.1 - 13 1434
          AK025648.1 - 7 648
          AK289674.1 - 13 1445
          AK292459.1 - 13 1502
          AK294850.1 - 12 1433
          BC036012.1 - 13 1466 5270430
          BX537955.1 - 14 1525

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          negative regulation of NF-kappaB transcription factor activity
          histone H2A monoubiquitination
          histone H2A-K119 monoubiquitination
          negative regulation of centriole replication
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of NF-kappaB transcription factor activity
          protein autoubiquitination
          aggresome assembly
          chromatin binding
          ubiquitin-protein transferase activity
          tumor necrosis factor receptor binding
          protein binding
          zinc ion binding
          ligase activity
          ubiquitin protein ligase binding
          protein homodimerization activity
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0