Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PUM2 SIRNA Assays ›
Gene Symbol:
PUM2
Gene Name:
pumilio RNA binding family member 2
Gene Aliases:
PUMH2, PUML2
Chromosome Location:
Chr.2: 20248692 - 20352423 on Build GRCh38
UniGene ID:
Hs.467824
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22867
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PUM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282752.1
NP_001269681.1
20
5395
NM_001282790.1
NP_001269719.1
19
5021
NM_001282791.1
NP_001269720.1
20
5146
NM_015317.2
NP_056132.1
21
5504
XM_005262607.1
XP_005262664.1
21
5484
XM_005262609.1
XP_005262666.1
20
5247
XM_005262610.1
XP_005262667.1
20
5241
XM_006711973.1
XP_006712036.1
21
5281
XM_006711974.2
XP_006712037.1
21
5321
XM_006711975.2
XP_006712038.1
22
5445
XM_011532719.1
XP_011531021.1
23
5680
XM_011532720.2
XP_011531022.1
23
5689
XM_011532721.2
XP_011531023.1
22
5481
XM_011532723.1
XP_011531025.1
20
5111
XM_017003701.1
XP_016859190.1
21
5275
XM_017003702.1
XP_016859191.1
21
5315
XM_017003703.1
XP_016859192.1
22
5509
XM_017003704.1
XP_016859193.1
24
5827
XM_017003705.1
XP_016859194.1
24
5814
XM_017003706.1
XP_016859195.1
23
5606
XM_017003707.1
XP_016859196.1
20
5044
XM_017003708.1
XP_016859197.1
20
5277
XM_017003709.1
XP_016859198.1
21
5237
XM_017003710.1
XP_016859199.1
20
5087
XM_017003711.1
XP_016859200.1
20
5038
XM_017003712.1
XP_016859201.1
20
5106
XM_017003713.1
XP_016859202.1
21
5265
XM_017003714.1
XP_016859203.1
17
4884
XM_017003715.1
XP_016859204.1
19
4868
GenBank mRNA
AF315591.1
-
20
5255
BX641165.1
-
1
4199
D87078.3
-
20
5232
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
posttranscriptional regulation of gene expression
stress granule assembly
regulation of mRNA stability
regulation of chromosome segregation
regulation of gene silencing by miRNA
positive regulation of RIG-I signaling pathway
positive regulation of gene silencing by miRNA
RNA binding
mRNA 3'-UTR binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-p8qt9:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0