Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090461.1 | - | 3 | 1282 | ||
AK126280.1 | - | 4 | 633 | |||
AK128568.1 | - | 5 | 651 | |||
AK128580.1 | - | 4 | 633 | |||
AK128614.1 | - | 4 | 690 | |||
AK128625.1 | - | 4 | 700 | |||
AK128635.1 | - | 4 | 711 | |||
AK128649.1 | - | 3 | 618 | |||
AK128658.1 | - | 3 | 709 | |||
BC021276.2 | - | 4 | 659 | |||
BC063384.1 | - | 4 | 620 |