siRNA Details
Gene Symbol: ANO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025356.2 | NP_001020527.2 | 4 | 634 | ||
NM_001142678.1 | NP_001136150.1 | 4 | 481 | |||
NM_001142679.1 | NP_001136151.1 | 4 | 634 | |||
NM_001204803.1 | NP_001191732.1 | 5 | 697 | |||
XM_005268706.4 | XP_005268763.1 | 4 | 588 | |||
XM_005268707.3 | XP_005268764.1 | 3 | 508 | |||
GenBank mRNA | AK126409.1 | - | 3 | 554 | ||
AK302819.1 | - | 5 | 491 | |||
AK303996.1 | - | 4 | 565 | |||
AK310641.1 | - | 4 | 601 | |||
AL832340.1 | - | 4 | 481 | |||
BC098410.1 | - | 4 | 501 | 30531341 | ||
BC136445.1 | - | 5 | 390 | 9020434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
