Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130969.1 | NP_001124441.1 | 9, 10 | 1281 | ||
NM_001130970.1 | NP_001124442.1 | 8, 9 | 1212 | |||
NM_001130971.1 | NP_001124443.1 | 7, 8 | 1206 | |||
NM_001178064.1 | NP_001171535.1 | 8, 9 | 1191 | |||
NM_015537.4 | NP_056352.3 | 8, 9 | 1275 | |||
XM_005266061.4 | XP_005266118.1 | 7, 8 | 1208 | |||
XM_005266062.4 | XP_005266119.1 | 6, 7 | 1139 | |||
XM_011518496.2 | XP_011516798.1 | 7, 8 | 1145 | |||
XM_011518497.2 | XP_011516799.1 | 4, 5 | 395 | |||
XM_017014597.1 | XP_016870086.1 | 3, 4 | 664 | |||
GenBank mRNA | AJ420418.1 | - | 1, 2 | 56 | ||
AK027474.1 | - | 6, 7 | 546 | |||
AK074602.1 | - | 8, 9 | 1243 | |||
AK292030.1 | - | 4, 5 | 347 | |||
AK294320.1 | - | 4, 5 | 324 | |||
AK303703.1 | - | 6, 7 | 403 | |||
AK303976.1 | - | 4, 5 | 442 | |||
AK316046.1 | - | 6, 7 | 597 | |||
AL117660.2 | - | 4, 5 | 303 | |||
AL389944.1 | - | 4, 5 | 329 | |||
AY255128.1 | - | 9, 10 | 1281 | |||
AY255129.1 | - | 8, 9 | 1275 | |||
AY255130.1 | - | 8, 9 | 1212 | |||
AY255131.1 | - | 8, 9 | 1191 | |||
BC016862.2 | - | 4, 5 | 322 | |||
BC072412.1 | - | 8, 9 | 1260 | 6165009 | ||
BC110498.2 | - | 7, 8 | 1073 | 40007676 |