Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MARK3 SIRNA Assays ›
          Gene Symbol:
          MARK3
          Gene Name:
          microtubule affinity regulating kinase 3
          Gene Aliases:
          CTAK1, KP78, PAR1A, Par-1a
          Chromosome Location:
          Chr.14: 103385364 - 103503831 on Build GRCh38
          UniGene ID:
          Hs.35828
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 230

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MARK3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001128918.2 NP_001122390.2 5, 6 1086
          NM_001128919.2 NP_001122391.2 5, 6 1086
          NM_001128920.2 NP_001122392.2 5, 6 1086
          NM_001128921.2 NP_001122393.2 5, 6 1086
          NM_002376.6 NP_002367.5 5, 6 1086
          XM_005267641.4 XP_005267698.2 5, 6 465
          XM_005267642.4 XP_005267699.2 5, 6 1086
          XM_005267643.4 XP_005267700.2 5, 6 1086
          XM_006720146.3 XP_006720209.1 5, 6 460
          XM_011536769.2 XP_011535071.1 4, 5 199
          XM_017021291.1 XP_016876780.1 5, 6 1086
          XM_017021292.1 XP_016876781.1 5, 6 1086
          XM_017021293.1 XP_016876782.1 6, 7 561
          XM_017021294.1 XP_016876783.1 4, 5 835
          XM_017021295.1 XP_016876784.1 4, 5 850
          XM_017021296.1 XP_016876785.1 4, 5 867
          XM_017021297.1 XP_016876786.1 4, 5 843
          XM_017021298.1 XP_016876787.1 4, 5 843
          XM_017021299.1 XP_016876788.1 4, 5 894
          XM_017021300.1 XP_016876789.1 3, 4 320
          XM_017021301.1 XP_016876790.1 3, 4 475
          XM_017021302.1 XP_016876791.1 3, 4 320
          XM_017021303.1 XP_016876792.1 3, 4 320
          XM_017021304.1 XP_016876793.1 3, 4 320
          XM_017021305.1 XP_016876794.1 3, 4 320
          XM_017021308.1 XP_016876797.1 4, 5 1037
          GenBank mRNA AF387637.1 - 5, 6 438
          AK307448.1 - 5, 6 465
          BC024773.1 - 5, 6 909 5138958
          BX161395.1 - 5, 6 1016
          U64205.1 - 5, 6 795

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_130749, XM_006240563, XM_006240568, XM_006240566, XM_006240571, and others.
          This siRNA also targets the following transcripts in Mouse: NM_021516, NM_022801, AK083683, AF240782, AF240783, and others.

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          microtubule cytoskeleton organization
          establishment of cell polarity
          protein serine/threonine kinase activity
          protein binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4j5fk:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0