Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080467.2 | NP_001073936.1 | 40 | 5788 | ||
GenBank mRNA | AB032945.2 | - | 40 | 5518 | ||
AB290160.1 | - | 40 | 5608 | |||
AK025336.1 | - | 19 | 2853 | |||
AK093149.1 | - | 18 | 2468 | |||
AK301976.1 | - | 12 | 1939 | |||
AL133643.1 | - | 18 | 2487 | |||
AY274809.1 | - | 18 | 2467 | |||
BC033527.1 | - | 9 | 1426 |