Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252402.2 | NP_001239331.1 | 5 | 709 | ||
NM_001252404.2 | NP_001239333.1 | 4 | 614 | |||
NM_001280561.1 | NP_001267490.1 | 6 | 1313 | |||
NR_047684.2 | 5 | 711 | ||||
GenBank mRNA | AB196634.1 | - | 6 | 1313 | ||
BC127763.1 | - | 4 | 528 | |||
BC127764.1 | - | 4 | 525 | |||
BC141966.1 | - | 5 | 624 | |||
BC150569.1 | - | 4 | 528 | |||
BC150570.1 | - | 4 | 528 | |||
BX648457.1 | - | 2 | 470 |