Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002774.3 | 1 | 1620 | |||
GenBank mRNA | AK125093.1 | - | 1 | 1628 | ||
U86813.1 | - | LIT2067 serotonin-7 receptor pseudogene |
1 | 560 |