siRNA Details
Gene Symbol: DYNC2H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080463.1 | NP_001073932.1 | 2 | 451 | ||
NM_001377.2 | NP_001368.2 | 2 | 451 | |||
XM_006718903.2 | XP_006718966.1 | 2 | 451 | |||
XM_017018291.1 | XP_016873780.1 | 2 | 451 | |||
XM_017018293.1 | XP_016873782.1 | 2 | 451 | |||
GenBank mRNA | AB231766.1 | - | 2 | 360 | ||
AB290167.1 | - | 2 | 505 | |||
AK125524.1 | - | 2 | 451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
