Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSIG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013661.1 | NP_001013683.1 | 3, 4 | 560 | ||
GenBank mRNA | BC132893.1 | - | 3, 4 | 452 | 40146915 | |
BC132895.1 | - | 3, 4 | 452 | 40146917 |