Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADARB1 SIRNA Assays ›
Gene Symbol:
ADARB1
Gene Name:
adenosine deaminase, RNA specific B1
Gene Aliases:
ADAR2, DRABA2, DRADA2, RED1
Chromosome Location:
Chr.21: 45074578 - 45226563 on Build GRCh38
UniGene ID:
Hs.474018
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
23261
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADARB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001112.3
NP_001103.1
4
785
NM_001160230.1
NP_001153702.1
4
785
NM_015833.3
NP_056648.1
4
785
NM_015834.3
NP_056649.1
4
785
NR_027673.1
5
903
NR_027674.1
4
785
NR_073200.1
4
785
XM_006723954.2
XP_006724017.1
5
857
XM_011529425.2
XP_011527727.1
5
1977
XM_017028242.1
XP_016883731.1
3
2453
XM_017028243.1
XP_016883732.1
6
5942
XM_017028244.1
XP_016883733.1
3
2451
XM_017028245.1
XP_016883734.1
3
2406
XM_017028246.1
XP_016883735.1
6
5942
XM_017028247.1
XP_016883736.1
5
5824
XM_017028248.1
XP_016883737.1
6
766
XM_017028249.1
XP_016883738.1
7
5985
XM_017028250.1
XP_016883739.1
6
5895
XM_017028251.1
XP_016883740.1
3
567
XM_017028252.1
XP_016883741.1
3
2453
XM_017028253.1
XP_016883742.1
3
2401
XM_017028254.1
XP_016883743.1
5
5824
XM_017028255.1
XP_016883744.1
5
5822
XM_017028256.1
XP_016883745.1
4
617
GenBank mRNA
AB194370.1
-
4
763
AB194371.1
-
4
763
AB194372.2
-
4
763
AB194373.1
-
4
763
BC065545.1
-
4
622
6014605
U76420.1
-
4
763
U76421.1
-
4
763
U76422.1
-
4
763
U82120.1
-
4
590
U82121.1
-
2
350
X99227.1
-
3
497
X99383.1
-
3
497
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
adenosine to inosine editing
mRNA processing
negative regulation of cell proliferation
negative regulation of cell migration
negative regulation of protein kinase activity by regulation of protein phosphorylation
positive regulation of viral genome replication
innate immune response
defense response to virus
regulation of cell cycle
double-stranded RNA adenosine deaminase activity
mRNA binding
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0