Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATP5S SIRNA Assays ›
          Gene Symbol:
          ATP5S
          Gene Name:
          ATP synthase, H+ transporting, mitochondrial Fo complex subunit s (factor B)
          Gene Aliases:
          ATPW, FB, HSU79253
          Chromosome Location:
          Chr.14: 50311752 - 50326715 on Build GRCh38
          UniGene ID:
          Hs.438489
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 23331

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ATP5S
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001003803.2 NP_001003803.1 3 951
          NM_001003805.2 NP_001003805.1 3 951
          NM_015684.3 NP_056499.2 3 951
          NR_033761.1 4 987
          XM_005267537.3 XP_005267594.1 3 858
          XM_011536649.2 XP_011534951.1 3 840
          XM_011536650.2 XP_011534952.1 3 842
          XM_011536651.2 XP_011534953.1 3 842
          XM_011536652.1 XP_011534954.1 3 240
          XM_011536653.2 XP_011534955.1 4 282
          XM_011536654.2 XP_011534956.1 3 846
          XM_011536655.2 XP_011534957.1 3 846
          XM_011536656.2 XP_011534958.1 3 878
          XM_011536657.2 XP_011534959.1 3 389
          XM_017021220.1 XP_016876709.1 3 322
          GenBank mRNA AF052186.1 - 3 903
          AF390029.1 - 2 175
          AK290008.1 - 3 639
          AY052377.1 - 3 374
          BC011549.1 - 3 951 4554461
          BC087851.1 - 4 756
          BG699167.1 - 3 287
          BU189283.1 - 3 262
          U79253.1 - 3 902

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ATP synthase primary active transporter transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          ATP biosynthetic process
          proton transport
          mitochondrial ATP synthesis coupled proton transport
          hydrogen ion transmembrane transporter activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-49ts5:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0