Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPKOW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015698.5 | NP_056513.2 | 2 | 288 | ||
XM_017029415.1 | XP_016884904.1 | 3 | 362 | |||
GenBank mRNA | AB209806.1 | - | 2 | 208 | ||
BC000397.2 | - | 2 | 216 | 2822421 | ||
BC003148.1 | - | 2 | 216 | 2822421 | ||
U66359.1 | - | 2 | 267 |