Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SETD5 SIRNA Assays ›
Gene Symbol:
SETD5
Gene Name:
SET domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.3: 9397700 - 9478154 on Build GRCh38
UniGene ID:
Hs.288164
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
233775
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SETD5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080517.2
NP_001073986.1
23
4935
NM_001292043.1
NP_001278972.1
25
5199
XM_005265301.1
XP_005265358.1
23
4973
XM_005265303.1
XP_005265360.1
26
5411
XM_011533920.1
XP_011532222.1
27
5506
XM_011533921.1
XP_011532223.1
26
5108
XM_011533922.1
XP_011532224.1
25
5069
XM_011533927.1
XP_011532229.1
27
5449
XM_011533928.1
XP_011532230.1
25
5012
XM_011533930.1
XP_011532232.1
27
5450
XM_011533932.1
XP_011532234.1
25
5217
XM_011533933.1
XP_011532235.1
23
4586
XM_017006767.1
XP_016862256.1
27
7168
XM_017006768.1
XP_016862257.1
25
6682
XM_017006769.1
XP_016862258.1
25
8785
XM_017006770.1
XP_016862259.1
25
8767
XM_017006771.1
XP_016862260.1
24
5030
XM_017006772.1
XP_016862261.1
24
8728
XM_017006773.1
XP_016862262.1
25
8728
XM_017006774.1
XP_016862263.1
24
4973
XM_017006775.1
XP_016862264.1
24
8671
XM_017006776.1
XP_016862265.1
26
5313
XM_017006777.1
XP_016862266.1
26
5256
XM_017006778.1
XP_016862267.1
27
5297
XM_017006779.1
XP_016862268.1
25
5274
XM_017006780.1
XP_016862269.1
26
5258
XM_017006781.1
XP_016862270.1
26
5240
XM_017006783.1
XP_016862272.1
28
5522
GenBank mRNA
AB051544.1
-
19
4729
AK001569.1
-
12
3118
AK024384.1
-
11
3029
AK025478.1
-
7
2043
AK091393.1
-
20
4238
AK094975.1
-
1
2687
AK127298.1
-
13
3822
AL442073.1
-
10
2915
BC020956.1
-
12
3149
3851178
CR627408.1
-
22
4551
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-tvqvm:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0