siRNA Details
Gene Symbol: AGPAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037553.1 | NP_001032642.1 | 9 | 5259 | ||
NM_020132.4 | NP_064517.1 | 10 | 5379 | |||
XM_005261160.4 | XP_005261217.1 | 10 | 5380 | |||
XM_006724029.3 | XP_006724092.1 | 9 | 5308 | |||
XM_006724030.3 | XP_006724093.1 | 9 | 5276 | |||
XM_006724031.3 | XP_006724094.1 | 9 | 5279 | |||
XM_011529660.2 | XP_011527962.1 | 9 | 5118 | |||
XM_011529661.2 | XP_011527963.1 | 9 | 5433 | |||
XM_011529662.2 | XP_011527964.1 | 11 | 6083 | |||
XM_011529663.2 | XP_011527965.1 | 9 | 5292 | |||
XM_011529664.2 | XP_011527966.1 | 9 | 5233 | |||
XM_011529665.2 | XP_011527967.1 | 11 | 5734 | |||
XM_017028408.1 | XP_016883897.1 | 9 | 5262 | |||
XM_017028409.1 | XP_016883898.1 | 9 | 5340 | |||
XM_017028410.1 | XP_016883899.1 | 9 | 5345 | |||
XM_017028411.1 | XP_016883900.1 | 9 | 5218 | |||
XM_017028412.1 | XP_016883901.1 | 10 | 6311 | |||
XM_017028413.1 | XP_016883902.1 | 10 | 5279 | |||
GenBank mRNA | AK074300.1 | - | 1 | 1302 | ||
AL355720.1 | - | 1 | 1575 | |||
AL355721.1 | - | 1 | 1102 | |||
AL832919.1 | - | 7 | 4953 | |||
BC004219.2 | - | 1 | 1438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
