Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FITM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080472.2 | NP_001073941.1 | 2 | 553 | ||
GenBank mRNA | BC130446.1 | - | 2 | 482 | 40146477 | |
BC136640.1 | - | 2 | 482 | 9020630 |