Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTNAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033655.3 | NP_387504.2 | 9 | 1695 | ||
GenBank mRNA | AB051501.2 | - | 9 | 1602 | ||
AF333769.2 | - | 9 | 1563 | |||
AK056833.1 | - | 9 | 1695 | |||
AK294281.1 | - | 9 | 1487 | |||
BC132737.1 | - | 9 | 1600 | 40146759 | ||
BC144130.1 | - | 9 | 1600 |
Gene Symbol: CNTNAP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201380.2 | NP_001188309.2 | 9 | 1866 |
Gene Symbol: CNTNAP3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111893.2 | 9 | 1555 | |||
GenBank mRNA | AK054645.1 | - | 9 | 1719 |
Gene Symbol: LOC100289279 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545672.2 | XP_011543974.1 | 9 | 1832 | ||
XM_017015380.1 | XP_016870869.1 | 9 | 1832 |