Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TAZ SIRNA Assays ›
Gene Symbol:
TAZ
Gene Name:
tafazzin
Gene Aliases:
BTHS, CMD3A, EFE, EFE2, G4.5, LVNCX, Taz1
Chromosome Location:
Chr.X: 154411406 - 154421726 on Build GRCh38
UniGene ID:
Hs.409911
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
235621
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TAZ
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000116.4
NP_000107.1
3
601
NM_001303465.1
NP_001290394.1
3
655
NM_181311.3
NP_851828.1
3
601
NM_181312.3
NP_851829.1
3
601
NM_181313.3
NP_851830.1
3
601
NR_024048.2
3
601
XM_006724836.1
XP_006724899.1
3
632
XM_006724837.1
XP_006724900.1
3
632
XM_006724839.1
XP_006724902.1
3
632
XM_006724841.3
XP_006724904.1
3
266
XM_006724842.3
XP_006724905.1
3
266
XM_011531189.1
XP_011529491.1
3
632
XM_011531191.2
XP_011529493.1
2
791
XM_017029761.1
XP_016885250.1
3
578
XM_017029762.1
XP_016885251.1
3
632
XM_017029763.1
XP_016885252.1
3
578
XM_017029764.1
XP_016885253.1
1
12
XM_017029765.1
XP_016885254.1
3
713
GenBank mRNA
AK291848.1
-
3
469
AL531401.3
-
3
603
AY231461.1
-
3
283
AY231462.1
-
3
283
AY231463.1
-
3
283
AY231464.1
-
3
283
AY258036.1
-
3
283
AY258037.1
-
3
283
AY258038.1
-
3
283
AY258039.1
-
3
283
BC005062.1
-
3
558
BC011515.1
-
2
297
4339983
BI822925.1
-
595
BX401320.2
-
3
441
CA488175.1
-
2
91
DQ884408.1
-
3
329
KU178348.1
-
2
200
KU178349.1
-
3
275
KU178350.1
-
3
275
KU178351.1
-
3
275
KU178352.1
-
3
275
KU178353.1
-
3
275
X92762.1
-
3
563
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001025748, XM_006229600, XM_006229599, BC099221.
This siRNA also targets the following transcripts in Mouse: NM_001242616, NM_001173547, NM_001242615, NM_001290738, NM_181516, and others.
Panther Classification:
Molecular Function -
acyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
heart development
skeletal muscle tissue development
hemopoiesis
cardiolipin biosynthetic process
mitochondrial respiratory chain complex I assembly
cardiolipin acyl-chain remodeling
cristae formation
mitochondrial ATP synthesis coupled electron transport
cardiac muscle tissue development
cardiac muscle contraction
1-acylglycerol-3-phosphate O-acyltransferase activity
O-acyltransferase activity
1-acylglycerophosphocholine O-acyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0