Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SSUH2 SIRNA Assays ›
          Gene Symbol:
          SSUH2
          Gene Name:
          ssu-2 homolog (C. elegans)
          Gene Aliases:
          C3orf32, SSU-2, fls485
          Chromosome Location:
          Chr.3: 8619400 - 8745035 on Build GRCh38
          UniGene ID:
          Hs.736935
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 23578

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SSUH2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256748.1 NP_001243677.1 6 504
          NM_001256749.1 NP_001243678.1 6 525
          NM_015931.2 NP_057015.1 6 671
          NR_046358.1 7 752
          XM_011533774.1 XP_011532076.1 14 2220
          XM_017006510.1 XP_016861999.1 7 4985
          XM_017006511.1 XP_016862000.1 13 1908
          XM_017006512.1 XP_016862001.1 13 2077
          XM_017006513.1 XP_016862002.1 12 1913
          XM_017006514.1 XP_016862003.1 12 1819
          XM_017006515.1 XP_016862004.1 14 2974
          XM_017006516.1 XP_016862005.1 9 1335
          XM_017006517.1 XP_016862006.1 9 1288
          XM_017006518.1 XP_016862007.1 14 2199
          XM_017006519.1 XP_016862008.1 7 4930
          XM_017006520.1 XP_016862009.1 7 1281
          XM_017006521.1 XP_016862010.1 10 1439
          XM_017006522.1 XP_016862011.1 10 1330
          XM_017006523.1 XP_016862012.1 8 1033
          XM_017006524.1 XP_016862013.1 9 1271
          XM_017006525.1 XP_016862014.1 12 1840
          XM_017006526.1 XP_016862015.1 7 919
          XM_017006527.1 XP_016862016.1 13 1962
          XM_017006528.1 XP_016862017.1 7 958
          XM_017006529.1 XP_016862018.1 7 988
          XM_017006530.1 XP_016862019.1 9 1421
          XM_017006531.1 XP_016862020.1 8 1343
          XM_017006532.1 XP_016862021.1 7 5843
          XM_017006533.1 XP_016862022.1 13 1995
          XM_017006534.1 XP_016862023.1 13 1941
          XM_017006535.1 XP_016862024.1 7 6119
          XM_017006536.1 XP_016862025.1 7 6122
          XM_017006537.1 XP_016862026.1 7 6122
          XM_017006538.1 XP_016862027.1 11 1538
          XM_017006539.1 XP_016862028.1 6 650
          XM_017006540.1 XP_016862029.1 7 829
          XM_017006541.1 XP_016862030.1 4 471
          XM_017006542.1 XP_016862031.1 4 449
          GenBank mRNA AB024705.1 - 6 644
          AK057972.1 - 7 748
          AK093066.1 - 12 1701
          AK122938.1 - 11 1486
          AK225925.1 - 7 748
          AK300175.1 - 6 504
          BC052614.1 - 6 508 6300057
          DB010779.1 - 6 525

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0