Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282874.1 | NP_001269803.1 | 7 | 987 | ||
NM_001282875.1 | NP_001269804.1 | 7 | 987 | |||
NM_003069.4 | NP_003060.2 | 7 | 987 | |||
XM_005262461.2 | XP_005262518.1 | 7 | 1002 | |||
XM_005262462.2 | XP_005262519.1 | 7 | 1005 | |||
XM_006724782.2 | XP_006724845.1 | 7 | 1046 | |||
XM_017029750.1 | XP_016885239.1 | 7 | 1046 | |||
XM_017029751.1 | XP_016885240.1 | 7 | 1046 | |||
GenBank mRNA | AK026426.1 | - | 7 | 862 | ||
BC051825.1 | - | 6 | 664 | |||
BC117447.1 | - | 7 | 887 | 40125998 | ||
BC143954.1 | - | 7 | 887 | 9052472 | ||
EU636009.1 | - | 7 | 946 | |||
M89907.1 | - | 7 | 1275 |