Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FASLG SIRNA Assays ›
          Gene Symbol:
          FASLG
          Gene Name:
          Fas ligand
          Gene Aliases:
          ALPS1B, APT1LG1, APTL, CD178, CD95-L, CD95L, FASL, TNFSF6, TNLG1A
          Chromosome Location:
          Chr.1: 172659008 - 172666873 on Build GRCh38
          UniGene ID:
          Hs.2007
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 2358

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FASLG
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000639.2 NP_000630.1 3, 4 653
          NM_001302746.1 NP_001289675.1 2, 3 607
          GenBank mRNA AF288573.1 - 2, 3 471
          AK314874.1 - 3, 4 617
          AY225406.1 - 3, 4 459
          AY858799.1 - 3, 4 459
          BC017502.1 - 3, 4 546 4849770
          D38122.1 - 3, 4 647
          KJ948655.1 - 3, 4 616
          U08137.1 - 3, 4 544
          U11821.1 - 3, 4 523
          X89102.1 - 3, 4 616

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          inflammatory cell apoptotic process
          immune response
          signal transduction
          cell-cell signaling
          positive regulation of cell proliferation
          extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of angiogenesis
          cellular chloride ion homeostasis
          response to lipopolysaccharide
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of neuron apoptotic process
          positive regulation of epidermal growth factor receptor signaling pathway
          retinal cell programmed cell death
          endosomal lumen acidification
          T cell apoptotic process
          necroptotic process
          response to growth factor
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          necroptotic signaling pathway
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          positive regulation of endothelial cell apoptotic process
          receptor binding
          death receptor binding
          cytokine activity
          tumor necrosis factor receptor binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0