siRNA Details
Gene Symbol: HOXB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018952.4 | NP_061825.2 | 4 | 1569 | ||
XM_005257283.3 | XP_005257340.1 | 3 | 1811 | |||
XM_005257284.3 | XP_005257341.1 | 3 | 1867 | |||
XM_011524727.2 | XP_011523029.1 | 4 | 2065 | |||
GenBank mRNA | AK292200.1 | - | 3 | 1867 | ||
BC014651.1 | - | 4 | 1569 | 4548382 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
